2-Bromobenzaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: o-Bromobenzaldehyde
OR13136-100G OR13136-25G
APOSOR13136-100GEA 24.8 EUR
APOSOR13136-100G APOSOR13136-25G
2-Bromobenzaldehyde
2-Bromobenzaldehyde
Formula: C₇H₅BrO
MW: 185.02 g/mol
Boiling Pt: 228…230 °C
Melting Pt: 16…21 °C
Density: 1.585 g/cm³ (25 °C)
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00003300
CAS Number: 6630-33-7
EINECS: 229-622-2