N-(4-Pyridyl)thiourea
Supplier: Apollo Scientific
Danger
OR1315-1G OR1315-5G
APOSOR1315-1GEA 46.4 EUR
APOSOR1315-1G APOSOR1315-5G
N-(4-Pyridyl)thiourea
N-(4-Pyridyl)thiourea
Formula: C₆H₇N₃S
Melting Pt: 181…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00068177
CAS Number: 164670-44-4
UN: 2811
ADR: 6.1,III