4-Hydroxybenzaldehyde
Supplier: Apollo Scientific
Warning
OR13187-1KG OR13187-500G OR13187-5KG
APOSOR13187-1KGEA 46.4 EUR
APOSOR13187-1KG APOSOR13187-500G APOSOR13187-5KG
4-Hydroxybenzaldehyde
4-Hydroxybenzaldehyde
Formula: C₇H₆O₂
MW: 122.12 g/mol
Boiling Pt: 191 °C/50mmHg
Melting Pt: 112…116 °C
Density: 1.129 g/cm³ (25 °C)
Flash Pt: 174 °C
Storage Temperature: Ambient
MDL Number: MFCD00006939
CAS Number: 123-08-0
EINECS: 204-599-1
Merck Index: 13,04836