1,1,3,3-Tetramethoxypropane 97%
Supplier: Apollo Scientific
Warning
Synonyms: Malonaldehyde bis(dimethyl acetal)
OR13192-1KG OR13192-25G OR13192-100G OR13192-500G
APOSOR13192-1KGEA 300 EUR
APOSOR13192-1KG APOSOR13192-25G APOSOR13192-100G APOSOR13192-500G
1,1,3,3-Tetramethoxypropane 97%
1,1,3,3-Tetramethoxypropane
Formula: C₇H₁₆O₄
MW: 164.2 g/mol
Boiling Pt: 182…183 °C
Density: 0.997 g/cm³ (25 °C)
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00008488
CAS Number: 102-52-3
EINECS: 203-037-2
UN: 1993
ADR: 3 III