3-Chloro-4-methylbenzaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 3-(5,6,6-Trimethyl-2-norbornyl)-cyclohexanol, isobornylcyclohexanol, 2-Chloro-4-formyltoluene
OR13197-1G OR13197-25G OR13197-5G
APOSOR13197-1GEA 0 EUR
APOSOR13197-1G APOSOR13197-25G APOSOR13197-5G
3-Chloro-4-methylbenzaldehyde
3-Chloro-4-methylbenzaldehyde
Formula: C₈H₇ClO
MW: 154.593 g/mol
Boiling Pt: 232 °C
Melting Pt: 27…31 °C
Storage Temperature: Ambient
MDL Number: MFCD06411284
CAS Number: 3411-03-8