Calcium nitrate tetrahydrate
Supplier: Apollo Scientific
Danger
APOSOR13216-1KGEA 51 EUR
APOSOR13216-1KG APOSOR13216-100G APOSOR13216-2.5KG
Calcium nitrate tetrahydrate
Calcium nitrate tetrahydrate
Formula: Ca(NO₃)₂·4H₂O
MW: 236,15 g/mol
Boiling Pt: 132 °C
Melting Pt: 40…45 °C
Density: 1,82 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149604
EINECS: 233-332-1
UN: 1454
ADR: 5.1,III