1,2-Dibromobenzene 98%
Supplier: Apollo Scientific
Warning
OR13221-100G OR13221-250G OR13221-25G
APOSOR13221-100GEA 53.8 EUR
APOSOR13221-100G APOSOR13221-250G APOSOR13221-25G
1,2-Dibromobenzene 98%
1,2-Dibromobenzene
Formula: C₆H₄Br₂
MW: 235.91 g/mol
Boiling Pt: 224 °C
Melting Pt: 44351 °C
Density: 1.956 g/cm³ (25 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00000057
CAS Number: 583-53-9
EINECS: 209-507-3