Ethyl-2-bromothiazole-5-carboxylate 97%
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 2-bromothiazole-5-carboxylate
OR1341-25G
APOSOR1341-25GEA 87 EUR
APOSOR1341-25G
Ethyl-2-bromothiazole-5-carboxylate 97%
Ethyl-2-bromothiazole-5-carboxylate
Formula: C₆H₆BrNO₂S
MW: 236.08 g/mol
Boiling Pt: 216…217 °C
Density: 1.581 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00463837
CAS Number: 41731-83-3