Indole-6-carbonitrile
Supplier: Apollo Scientific
Warning
Synonyms: 6-Cyano-1H-indole, 6-Cyanoindole, 1H-Indole-6-carbonitrile
OR1347-5G OR1347-10G
APOSOR1347-5GEA 21.9 EUR
APOSOR1347-5G APOSOR1347-10G
Indole-6-carbonitrile
Indole-6-carbonitrile
Formula: C₉H₆N₂
MW: 142.16 g/mol
Melting Pt: 128…132 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00016732
CAS Number: 15861-36-6
EINECS: 239-988-5