6-Methyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-sulphonyl chloride 95%
Supplier: Apollo Scientific
Danger
Synonyms: 6-Methyluracil-5-sulphonyl chloride
OR13481-1G
APOSOR13481-1GEA 516 EUR
APOSOR13481-1G
6-Methyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-sulphonyl chloride 95%
6-Methyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-sulphonyl chloride
Formula: C₅H₅ClN₂O₄S
MW: 224.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00476725
CAS Number: 6461-30-9
UN: 1759
ADR: 8,III