1,2-Dimethyl-1,4,5,6-tetrahydropyrimidine
Supplier: Apollo Scientific
Danger
OR13592-100G OR13592-25G OR13592-500G
APOSOR13592-100GEA 31.9 EUR
APOSOR13592-100G APOSOR13592-25G APOSOR13592-500G
1,2-Dimethyl-1,4,5,6-tetrahydropyrimidine
1,2-Dimethyl-1,4,5,6-tetrahydropyrimidine
Formula: C₆H₁₂N₂
Boiling Pt: 125 °C
Density: 0.98 g/cm³ (25 °C)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00036056
CAS Number: 4271-96-9
EINECS: 224-262-2
UN: 3267
ADR: 8,III