2-Acetyl-6-methylpyridine
Supplier: Apollo Scientific
Warning
OR13601-1G OR13601-5G OR13601-25G
APOSOR13601-1GEA 21.9 EUR
APOSOR13601-1G APOSOR13601-5G APOSOR13601-25G
2-Acetyl-6-methylpyridine
2-Acetyl-6-methylpyridine
Formula: C₈H₉NO
MW: 135.17 g/mol
Boiling Pt: 209 °C
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD06658292
CAS Number: 6940-57-4