2-Chloro-3-picoline 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Chloro-3-methylpyridine, 2-Chloro 3-methylpyridine, 2-Chloro-β-picoline
OR13603-500G
APOSOR13603-500GEA 98.6 EUR
APOSOR13603-500G
2-Chloro-3-picoline 97%
2-Chloro-3-picoline
Formula: C₆H₆ClN
MW: 127.57 g/mol
Boiling Pt: 192…193 °C
Density: 1.17 g/cm³ (25 °C)
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00234177
CAS Number: 18368-76-8
EINECS: 242-242-1