4-(2-Hydroxyethyl)pyridine 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2-(4-Pyridyl)ethanol
OR13623-5G
APOSOR13623-5GEA 277 EUR
APOSOR13623-5G
4-(2-Hydroxyethyl)pyridine 97%
4-(2-Hydroxyethyl)pyridine
Formula: C₇H₉NO
Boiling Pt: 121…122 °C/2mmHg
Melting Pt: 44477 °C
Density: 1.093 g/cm³ (25 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00129038
CAS Number: 5344-27-4
EINECS: 226-286-9