2,6-Diaminopyridine
Supplier: Apollo Scientific
Warning
OR13639-100G OR13639-500G
APOSOR13639-100GEA 102 EUR
APOSOR13639-100G APOSOR13639-500G
2,6-Diaminopyridine
2,6-Diaminopyridine
Formula: C₅H₇N₃
MW: 109.13 g/mol
Boiling Pt: 285 °C
Melting Pt: 117…122 °C
Flash Pt: 155 °C
Storage Temperature: Ambient
MDL Number: MFCD00006329
CAS Number: 141-86-6
EINECS: 205-507-2
UN: 2811
ADR: 6.1,III