6-Chloro-o-toluidine 97%
Supplier: Apollo Scientific
OR13655-100G OR13655-25G OR13655-500G
APOSOR13655-100GEA 85.7 EUR
APOSOR13655-100G APOSOR13655-25G APOSOR13655-500G
6-Chloro-o-toluidine 97%
6-Chloro-o-toluidine
Formula: C₇H₈ClN
MW: 141.6 g/mol
Boiling Pt: 215 °C
Melting Pt: 44540 °C
Density: 1.152 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007679
CAS Number: 87-63-8