Tetrahydro-1,3-dimethyl-1H-pyrimidin-2-one
Supplier: Apollo Scientific
Danger
Synonyms: Dimethylpropyleneurea, Dimethyl propylene urea, 1,3-Dimethyl-3,4,5,6-tetrahydro-2(1H)-pyrimidinone, 1,3-Dimethyltetrahydropyrimidin-2(1H)-one, DMPU, 1,3-Dimethyltetrahydro-2(1H)-pyrimidinone, N,N'-Dimethyl propylene urea
OR13656-100G OR13656-25G
APOSOR13656-100GEA 24.8 EUR
APOSOR13656-100G APOSOR13656-25G
Tetrahydro-1,3-dimethyl-1H-pyrimidin-2-one
Tetrahydro-1,3-dimethyl-1H-pyrimidin-2-one
Formula: C₆H₁₂N₂O
MW: 128.17 g/mol
Boiling Pt: 246…247 °C
Melting Pt: –22.99 °C
Density: 1.06 g/cm³ (25 °C)
Flash Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00006550
CAS Number: 7226-23-5
EINECS: 230-625-6