2-Chlorobenzoxazole 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Chloro-1,3-benzoxazole
OR13659-5G OR13659-1G
APOSOR13659-5GEA 24.8 EUR
APOSOR13659-5G APOSOR13659-1G
2-Chlorobenzoxazole 97%
2-Chlorobenzoxazole
Formula: C₇H₄ClNO
MW: 153.57 g/mol
Boiling Pt: 201…202 °C
Melting Pt: 7 °C
Density: 1.345 g/cm³ (25 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00005766
CAS Number: 615-18-9
EINECS: 210-414-5