1,3-Bis(chloromethyl)tetramethyldisiloxane 95%
Supplier: Apollo Scientific
OR13693-100G OR13693-25G
APOSOR13693-100GEA 133 EUR
APOSOR13693-100G APOSOR13693-25G
1,3-Bis(chloromethyl)tetramethyldisiloxane 95%
1,3-Bis(chloromethyl)tetramethyldisiloxane
Formula: C₆H₁₆Cl₂OSi₂
Storage Temperature: Ambient
MDL Number: MFCD00000877
CAS Number: 2362-10-9