6-Aminohexan-1-ol 98%
Supplier: Apollo Scientific
Warning
Synonyms: 6-Amino-1-hexanol
OR13708-25G OR13708-5G
APOSOR13708-25GEA 24.8 EUR
APOSOR13708-25G APOSOR13708-5G
6-Aminohexan-1-ol 98%
6-Aminohexan-1-ol
Formula: C₆H₁₅NO
MW: 117.19 g/mol
Boiling Pt: 136…139 °C
Melting Pt: 55…58 °C
Flash Pt: 133 °C
Storage Temperature: Ambient
MDL Number: MFCD00008241
CAS Number: 4048-33-3