(R)-2-(Aminomethyl)-1-N-boc-pyrrolidine 96%
Supplier: Apollo Scientific
Danger
Synonyms: tert-Butyl (R)-2-(Aminomethyl)-1-pyrrolidinecarboxylate, (R)-2-(Aminomethyl)-1-Boc-pyrrolidine, (R)-2-(Aminomethyl)-1-(tert-butoxycarbonyl)pyrrolidine, (R)-2-(Aminomethyl)-1-pyrrolidinecarboxylic acid tert-Butyl ester
OR1380-1G OR1380-5G
APOSOR1380-1GEA 21.9 EUR
APOSOR1380-1G APOSOR1380-5G
(R)-2-(Aminomethyl)-1-N-boc-pyrrolidine 96%
(R)-2-(Aminomethyl)-1-N-boc-pyrrolidine
Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
Boiling Pt: 98…110 °C/1mmHg
Storage Temperature: Ambient
MDL Number: MFCD03419256
CAS Number: 259537-92-3
UN: 2735
ADR: 8 III