Pentaethylene glycol 97%
Supplier: Apollo Scientific
Warning
Synonyms: 3,6,9,12-Tetraoxatetradecane-1,14-diol
OR13897-10G OR13897-5G OR13897-25G
APOSOR13897-10GEA 79.9 EUR
APOSOR13897-10G APOSOR13897-5G APOSOR13897-25G
Pentaethylene glycol 97%
Pentaethylene glycol
Formula: C₁₀H₂₂O₆
MW: 238.28 g/mol
Boiling Pt: 338…340 °C
Density: 1.126 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002878
CAS Number: 4792-15-8
EINECS: 225-341-4