3-Thiopheneethanol 98%
Supplier: Apollo Scientific
Synonyms: Thiophen-3-ethanol, 2-(Thiophen-3-yl)ethanol, 2-(3-Thienyl)ethanol
OR13899-100G OR13899-25G
APOSOR13899-100GEA 422 EUR
APOSOR13899-100G APOSOR13899-25G
3-Thiopheneethanol 98%
3-Thiopheneethanol
Formula: C₆H₈OS
Boiling Pt: 110…111 °C/14mmHg
Density: 1.14 g/cm³ (25 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00009766
CAS Number: 13781-67-4