4-Cyano-4'-pentylbiphenyl 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4'-Pentyl-4-biphenylcarbonitrile, 4'-Pentyl-[1,1'-biphenyl]-4-carbonitrile
OR13991-5G OR13991-10G OR13991-1G
APOSOR13991-5GEA 84.1 EUR
APOSOR13991-5G APOSOR13991-10G APOSOR13991-1G
4-Cyano-4'-pentylbiphenyl 98%
4-Cyano-4'-pentylbiphenyl
Formula: C₁₈H₁₉N
MW: 249.36 g/mol
Melting Pt: 212…213/3mm °C
Density: 1.008 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00036350
CAS Number: 40817-08-1
EINECS: 255-093-2