2-Hydroxyquinoline-3-carbaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 2(1H)-Quinolinone-3-carboxaldehyde, 2-Oxo-1,2-dihydro-3-quinolinecarbaldehyde
OR14001-1G OR14001-5G
APOSOR14001-1GEA 105 EUR
APOSOR14001-1G APOSOR14001-5G
2-Hydroxyquinoline-3-carbaldehyde
2-Hydroxyquinoline-3-carbaldehyde
Formula: C₁₀H₇NO₂
MW: 173.171 g/mol
Melting Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD00563443
CAS Number: 91301-03-0