Ethyl-5-bromo-1-methyl-1H-indole-2-carboxylate
Supplier: Apollo Scientific
Warning
OR14054-1G OR14054-5G OR14054-250MG
APOSOR14054-1GEA 183 EUR
APOSOR14054-1G APOSOR14054-5G APOSOR14054-250MG
Ethyl-5-bromo-1-methyl-1H-indole-2-carboxylate
Ethyl-5-bromo-1-methyl-1H-indole-2-carboxylate
Formula: C₁₂H₁₂BrNO₂
MW: 282.14 g/mol
Melting Pt: 83…85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09027165
CAS Number: 91844-20-1