1-(3-Acetylphenyl)-2-thiourea
Supplier: Apollo Scientific
Warning
OR1414-10G OR1414-50G
APOSOR1414-10GEA 270 EUR
APOSOR1414-10G APOSOR1414-50G
1-(3-Acetylphenyl)-2-thiourea
1-(3-Acetylphenyl)-2-thiourea
Formula: C₉H₁₀N₂OS
Melting Pt: 165…167 °C
Storage Temperature: Ambient
MDL Number: MFCD00041170
CAS Number: 86801-04-9
UN: 3335
ADR: 9,III