Methyl-4,6-dichloronicotinate 95+%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 4,6-dichloronicotinate
OR14223-1G OR14223-25G OR14223-5G
APOSOR14223-1GEA 21.9 EUR
APOSOR14223-1G APOSOR14223-25G APOSOR14223-5G
Methyl-4,6-dichloronicotinate 95+%
Methyl 4,6-dichloronicotinate
Formula: C₇H₅Cl₂NO₂
MW: 206.03 g/mol
Melting Pt: 41…43 °C
Storage Temperature: Freezer
MDL Number: MFCD04125732
CAS Number: 65973-52-6