Methyl imidazo[1,2-a]pyridine-6-carboxylate
Supplier: Apollo Scientific
Warning
OR14226-1G OR14226-5G
APOSOR14226-1GEA 53.8 EUR
APOSOR14226-1G APOSOR14226-5G
Methyl imidazo[1,2-a]pyridine-6-carboxylate
Methyl imidazo[1,2-a]pyridine-6-carboxylate
Formula: C₉H₈N₂O₂
MW: 176.1720 g/mol
Melting Pt: 147…148 °C
Storage Temperature: Ambient
MDL Number: MFCD06200984
CAS Number: 136117-69-6