1-Boc-5-methoxyindole-2-boronic acid
Supplier: Apollo Scientific
Warning
Synonyms: N-Boc-5-methoxy-2-indolylboronic acid, (1-(tert-Butoxycarbonyl)-5-methoxy-1H-indol-2-yl)boronic acid
OR14256-1G OR14256-5G OR14256-25G
APOSOR14256-1GEA 68.3 EUR
APOSOR14256-1G APOSOR14256-5G APOSOR14256-25G
1-Boc-5-methoxyindole-2-boronic acid
1-Boc-5-methoxyindole-2-boronic acid
Formula: C₁₄H₁₈BNO₅
MW: 291.11 g/mol
Melting Pt: 102…108 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04039006
CAS Number: 290331-71-4