N-(3-Formylphenyl)methanesulphonamide 97%
Supplier: Apollo Scientific
Danger
Synonyms: 3-(Methylsulphonamido)benzaldehyde
OR14259-1G OR14259-5G OR14259-100MG
APOSOR14259-1GEA 145 EUR
APOSOR14259-1G APOSOR14259-5G APOSOR14259-100MG
N-(3-Formylphenyl)methanesulphonamide 97%
N-(3-Formylphenyl)methanesulphonamide
Formula: C₈H₉NO₃S
MW: 199.23 g/mol
Melting Pt: 146…147 °C
Storage Temperature: Ambient
MDL Number: MFCD03198179
CAS Number: 55512-05-5