Dess-Martin periodinane
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1,1-Dihydro-1,1,1-triacetoxy-1,2-benziodoxol-3(1H)-one, DMP, (1,1,1-Triacetoxy)-1,1-dihydro-1,2-benziodoxol-3(1H)-one, Acetic acid 1,1-diacetoxy-3-oxo-1l5-ioda-2-oxa-indan-1-yl ester
OR14263-25G
OR14263-100G
OR14263-250G
APOSOR14263-25GEA
29
EUR
APOSOR14263-25G
APOSOR14263-100G
APOSOR14263-250G
Dess-Martin periodinane
Dess-Martin periodinane
Formula:
C₁₃H₁₃IO₈ Melting Pt: 130…133 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00130127 CAS Number: 87413-09-0 UN: 1479 ADR: 5.1,III |