6-Hydroxy-2-naphthaldehyde 98%
Supplier: Apollo Scientific
Warning
OR14282-100G OR14282-25G OR14282-5G
APOSOR14282-100GEA 463 EUR
APOSOR14282-100G APOSOR14282-25G APOSOR14282-5G
6-Hydroxy-2-naphthaldehyde 98%
6-Hydroxy-2-naphthaldehyde
Formula: C₁₁H₈O₂
MW: 172.1800 g/mol
Melting Pt: 183 °C
Storage Temperature: Ambient
MDL Number: MFCD03427537
CAS Number: 78119-82-1