5-Phenyl-4-(1H-1,2,4-triazol-1-yl)-1,3-cyclohexanedione
Supplier: Apollo Scientific
Warning
OR14296-1G OR14296-500MG
APOSOR14296-1GEA 116 EUR
APOSOR14296-1G APOSOR14296-500MG
5-Phenyl-4-(1H-1,2,4-triazol-1-yl)-1,3-cyclohexanedione
5-Phenyl-4-(1H-1,2,4-triazol-1-yl)-1,3-cyclohexanedione
Formula: C₁₄H₁₃N₃O₂
MW: 255.28 g/mol
Melting Pt: 221…223 °C
Storage Temperature: Ambient
MDL Number: MFCD06411615
CAS Number: 886361-84-8