3-Bromoanthranilic acid 97%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Amino-3-bromobenzoic acid
OR14301-5G OR14301-100G OR14301-25G
APOSOR14301-5GEA 21.9 EUR
APOSOR14301-5G APOSOR14301-100G APOSOR14301-25G
3-Bromoanthranilic acid 97%
3-Bromoanthranilic acid
Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Boiling Pt: 343…344 °C
Melting Pt: 174…178 °C
Density: 1.793 g/cm³ (25 °C)
Flash Pt: 161 °C
Storage Temperature: Ambient
MDL Number: MFCD03618453
CAS Number: 20776-51-6
UN: 2811
ADR: 6.1 III