4-Hydroxytetrahydropyran 97%
Supplier: Apollo Scientific
Danger
Synonyms: Tetrahydro-2H-pyran-4-ol, Tetrahydro-4H-pyran-4-ol
OR14305-100G OR14305-25G OR14305-5G
APOSOR14305-100GEA 116 EUR
APOSOR14305-100G APOSOR14305-25G APOSOR14305-5G
4-Hydroxytetrahydropyran 97%
4-Hydroxytetrahydropyran
Formula: C₅H₁₀O₂
MW: 102.1317 g/mol
Boiling Pt: 87 °C/15mmHg
Density: 1.071 g/cm³ (25 °C)
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00006633
CAS Number: 2081-44-9
EINECS: 218-210-8