10-Hydroxydecanoic acid 98%
Supplier: Apollo Scientific
Warning
OR14308-25G OR14308-5G OR14308-100G
APOSOR14308-25GEA 79.9 EUR
APOSOR14308-25G APOSOR14308-5G APOSOR14308-100G
10-Hydroxydecanoic acid 98%
10-Hydroxydecanoic acid
Formula: C₁₀H₂₀O₃
MW: 188.26 g/mol
Melting Pt: 75…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00010510
CAS Number: 1679-53-4
EINECS: 216-848-1