4-Bromoisoquinoline
Supplier: Apollo Scientific
Warning
OR14319-25G OR14319-5G
APOSOR14319-25GEA 33.5 EUR
APOSOR14319-25G APOSOR14319-5G
4-Bromoisoquinoline
4-Bromoisoquinoline
Formula: C₉H₆BrN
MW: 208.06 g/mol
Boiling Pt: 280…285 °C
Melting Pt: 40…43 °C
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006904
CAS Number: 1532-97-4
EINECS: 216-244-8