N,N-Dimethyl-1,4-phenylendiamine hydrochloride
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
N1,N1-Dimethylbenzene-1,4-diamine hydrochloride, N,N-Dimethyl-p-phenylenediamine monohydrochloride
OR14326-25G
OR14326-5G
APOSOR14326-25GEA
116
EUR
APOSOR14326-25G
APOSOR14326-5G
N,N-Dimethyl-1,4-phenylendiamine hydrochloride
N,N-Dimethyl-1,4-phenylendiamine hydrochloride
Formula:
C₈H₁₃ClN₂ MW: 172.66 g/mol Melting Pt: 215 °C (dec.) Storage Temperature: Freezer |
MDL Number:
MFCD00012990 CAS Number: 2052-46-2 EINECS: 218-146-0 UN: 2811 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...