4-Methoxysalicylic acid
Supplier: Apollo Scientific
Warning
Synonyms: 2-Hydroxy-p-anisic acid, 2-Hydroxy-4-methoxybenzoic acid
OR14335-100G OR14335-25G
APOSOR14335-100GEA 79.9 EUR
APOSOR14335-100G APOSOR14335-25G
4-Methoxysalicylic acid
4-Methoxysalicylic acid
Formula: C₈H₈O₄
MW: 168.15 g/mol
Boiling Pt: 327
Melting Pt: 157…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00002450
CAS Number: 2237-36-7
EINECS: 218-801-0