Methyl-5-bromosalicylate
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 5-bromo-2-hydroxybenzoate
OR14417-25G OR14417-5G OR14417-100G
APOSOR14417-25GEA 48 EUR
APOSOR14417-25G APOSOR14417-5G APOSOR14417-100G
Methyl-5-bromosalicylate
Methyl 5-bromosalicylate
Formula: C₈H₇BrO₃
MW: 231.05 g/mol
Melting Pt: 61…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00016460
CAS Number: 4068-76-2
EINECS: 223-776-4