5-Chloro-1-pentyne 98%
Supplier: Apollo Scientific
Danger
OR14511-25G OR14511-5G
APOSOR14511-25GEA 120 EUR
APOSOR14511-25G APOSOR14511-5G
5-Chloro-1-pentyne 98%
5-Chloro-1-pentyne
Formula: C₅H₇Cl
MW: 102.56 g/mol
Boiling Pt: 67…69 °C/145mmHg
Density: 0.968 g/cm³ (25 °C)
Flash Pt: 15 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001014
CAS Number: 14267-92-6
EINECS: 238-159-5
UN: 1993
ADR: 3,II