N-Hydroxy-5-norbornene-2,3-dicarboximide 97%
Supplier: Apollo Scientific
Synonyms: endo-N-Hydroxybicyclo[2.2.1]hept-5-ene-2,3-dicarboximide
OR14538-25G OR14538-100G OR14538-500G
APOSOR14538-25GEA 43.7 EUR
APOSOR14538-25G APOSOR14538-100G APOSOR14538-500G
N-Hydroxy-5-norbornene-2,3-dicarboximide 97%
N-Hydroxy-5-norbornene-2,3-dicarboximide
Formula: C₉H₉NO₃
MW: 179.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00167583
CAS Number: 21715-90-2