Fmoc-3,4-dichloro-L-phenylalanine 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Fmoc-Phe(3,4-DiCl)-OH, (S)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(3,4-dichlorophenyl)propanoic acid, 3,4-Dichloro-L-phenylalanine, N-FMOC protected
OR14608-5G
OR14608-1G
APOSOR14608-5GEA
255
EUR
APOSOR14608-5G
APOSOR14608-1G
Fmoc-3,4-dichloro-L-phenylalanine 95%
Fmoc-3,4-dichloro-L-phenylalanine
Formula:
C₂₄H₁₉Cl₂NO₄ Melting Pt: 122 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00273473 CAS Number: 177966-59-5 |