Fmoc-4-iodo-L-phenylalanine 98%
Supplier: Apollo Scientific
Warning
Synonyms: Fmoc-p-iodo-Phe-OH, Fmoc-Phe(4-I)-OH
OR14612-1G OR14612-5G
APOSOR14612-1GEA 21.9 EUR
APOSOR14612-1G APOSOR14612-5G
Fmoc-4-iodo-L-phenylalanine 98%
Fmoc-4-iodo-L-phenylalanine
Formula: C₂₄H₂₀INO₄
MW: 513.331 g/mol
Melting Pt: 213…217 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00672340
CAS Number: 82565-68-2