3-(2-Naphthyl)-D-alanine
Supplier: Apollo Scientific
Warning
OR14714-1G OR14714-5G
APOSOR14714-1GEA 24.4 EUR
APOSOR14714-1G APOSOR14714-5G
3-(2-Naphthyl)-D-alanine
3-(2-Naphthyl)-D-alanine
Formula: C₁₃H₁₃NO₂
Melting Pt: 244…245 °C
Storage Temperature: Ambient
MDL Number: MFCD00038546
CAS Number: 76985-09-6