L-Lysine
Supplier: Apollo Scientific
Warning
Synonyms: (S)-2,6-Diaminohexanoic acid, L(+)-Lysine, (S)-(+)-2,6-Diaminohexanoic acid, (S)-(+)-Lysine, (S)-(+)-2,6-Diaminocaproic acid, H-Lys-OH
OR14752-100G OR14752-1KG OR14752-500G
APOSOR14752-100GEA 26.1 EUR
APOSOR14752-100G APOSOR14752-1KG APOSOR14752-500G
L-Lysine
L-Lysine
Formula: C₆H₁₄N₂O₂
MW: 146.19 g/mol
Melting Pt: 215 °C
Storage Temperature: Ambient
MDL Number: MFCD00064433
CAS Number: 56-87-1
EINECS: 200-294-2