3-Methyl-5-nitro-1H-pyrazole
Supplier: Apollo Scientific
Warning
OR14841-1G OR14841-5G OR14841-25G
APOSOR14841-1GEA 21.9 EUR
APOSOR14841-1G APOSOR14841-5G APOSOR14841-25G
3-Methyl-5-nitro-1H-pyrazole
3-Methyl-5-nitro-1H-pyrazole
Formula: C4H5N3O2
MW: 127.1014 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00463977
CAS Number: 34334-96-8