4-Methylquinoline
Supplier: Apollo Scientific
Warning
Synonyms: Lepidine
OR14907-100G OR14907-25G
APOSOR14907-100GEA 259 EUR
APOSOR14907-100G APOSOR14907-25G
4-Methylquinoline
4-Methylquinoline
Formula: C₁₀H₉N
MW: 143.19 g/mol
Boiling Pt: 261…263 °C
Melting Pt: 44478 °C
Density: 1.083 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00006784
CAS Number: 491-35-0
EINECS: 207-734-2
Merck Index: 13,05460