2,6-Dichloropyridine 98%
Supplier: Apollo Scientific
Danger
OR1491-100G OR1491-500G
APOSOR1491-100GEA 21.9 EUR
APOSOR1491-100G APOSOR1491-500G
2,6-Dichloropyridine 98%
2,6-Dichloropyridine
Formula: C₅H₃Cl₂N
MW: 147.99 g/mol
Boiling Pt: 211 °C
Melting Pt: 84…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00006244
CAS Number: 2402-78-0
EINECS: 219-282-3
UN: 2811
ADR: 6.1,III